Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401326

Sigma-Aldrich

Anti-RAGE antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MOK, RAGE1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAGE(5891)

Immunogen

RAGE (AAH26069.1, 1 a.a. ~ 237 a.a) full-length human protein.

Sequence
MDLWSAGCVFYEIASLQPLFPGVNELDQISKIHDVIGTPAQKILTKFKQSRAMNFDFPFKKGSGIPLLTTNLSPQCLSLLHAMVAYDPDERIAAHQALQHPYFQEQRKTEKRALGSHRKAGFPEHPVAPEPLSNSCQISKEGRKQKQSLKQEEDRPKRRGPAYVMELPKLKLSGVVRLSSYSSPTLQSVLGSGTNGRVPVLRPLKCIPASKKTDPQKDLKPAPQQCRLPTIVRKGGR

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Amalia Guzmán et al.
Investigational new drugs, 37(2), 262-270 (2018-07-13)
Pancreatic cancer, the fourth leading cause of cancer death in the United States, has a negative prognosis because metastasis occurs before symptoms manifest. Although combination therapies are showing improvements in treatment, the survival rate for pancreatic cancer five years post
Dong Zhao et al.
Molecular medicine reports, 14(6), 4983-4990 (2016-11-15)
Tanshinone II A (TSIIA) is a diterpene quinone extracted from the roots of Salvia miltiorrhiza with anti-inflammatory and anti‑oxidant properties that is used to treat atherosclerosis. In the current study, morphological analyses were conducted to evaluate the effects of TSIIA on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service