Skip to Content
Merck
All Photos(1)

Documents

S7065

Sigma-Aldrich

Secretoneurin mouse, rat trifluoroacetate salt

Synonym(s):

Secretogranin II (154-186)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C159H252N40O58
Molecular Weight:
3651.94
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.77

UniProt accession no.

Quality Level

storage temp.

−20°C

Gene Information

mouse ... Scg2(20254)
rat ... Scg2(24765)

Amino Acid Sequence

Thr-Asn-Glu-Ile-Val-Glu-Glu-Gln-Tyr-Thr-Pro-Gln-Ser-Leu-Ala-Thr-Leu-Glu-Ser-Val-Phe-Gln-Glu-Leu-Gly-Lys-Leu-Thr-Gly-Pro-Ser-Asn-Gln

Biochem/physiol Actions

Neuropeptide generated in brain, adrenal medulla and other endocrine tissues by proteolytic processing of secretogranin II (chromogranin C). Endogenous peptide that enhances dopamine release.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service