P4810
IgG2a−PE Isotype Control from murine myeloma
clone UPC-10, purified immunoglobulin, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
Recommended Products
biological source
mouse
conjugate
phycoerythrin (R-PE) conjugate
antibody form
purified immunoglobulin
clone
UPC-10, monoclonal
form
buffered aqueous solution
storage condition
protect from light
technique(s)
flow cytometry: suitable
shipped in
wet ice
storage temp.
2-8°C
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide
Preparation Note
Prepared by conjugation to R-phycoerythrin (PE). This orange-red dye is efficiently excited at 488 nm and emits at 578 nm.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Min Song et al.
Journal of experimental & clinical cancer research : CR, 31, 23-23 (2012-03-20)
Multidrug resistance is one of the main impediments to the successful treatment of colon cancer. Glucosylceramide synthase (GCS) which is related to multidrug resistance (MDR) can reduce the level of ceramide and can help cells escape from the ceramide-induced cell
H E Høgevold et al.
European surgical research. Europaische chirurgische Forschung. Recherches chirurgicales europeennes, 28(3), 190-200 (1996-01-01)
The aim of this study was to characterize the changes in the quantitative expression of beta 2-integrins and L-selectin detected by means of fluorochrome-conjugated monoclonal antibodies and flow cytometry on leukocytes in the systemic circulation after a major musculoskeletal trauma
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service