Skip to Content
Merck
All Photos(1)

Key Documents

P205

Supelco

Propentofylline

Synonym(s):

3,7-Dihydro-3-methyl-1-(5-oxohexyl)-7-propyl-1H-purine-2,6-dione, 3-Methyl-1-(5-oxohexyl)-7-propylxanthine, HWA 285

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N4O3
CAS Number:
Molecular Weight:
306.36
MDL number:
UNSPSC Code:
41116107

storage temp.

−20°C

SMILES string

CCCn1cnc2N(C)C(=O)N(CCCCC(C)=O)C(=O)c12

InChI

1S/C15H22N4O3/c1-4-8-18-10-16-13-12(18)14(21)19(15(22)17(13)3)9-6-5-7-11(2)20/h10H,4-9H2,1-3H3

InChI key

RBQOQRRFDPXAGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

c-AMP phosphodiesterase inhibitor; nerve growth factor stimulator; adenosine transport inhibitor; non-selective adenosine receptor antagonist.

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service