Skip to Content
Merck
All Photos(1)

Key Documents

K1888

Sigma-Aldrich

K 185

solid

Synonym(s):

N-Butanoyl 2-(5,6,7-trihydro-11-methoxybenzo[c]cyclohept­[2,1-a]­indol-13-yl)ethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28N2O2
CAS Number:
Molecular Weight:
376.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

DMSO: soluble 20 mg/mL

storage temp.

−20°C

SMILES string

CCCC(=O)NCCc1c2-c3ccccc3CCCn2c4ccc(OC)cc14

InChI

1S/C24H28N2O2/c1-3-7-23(27)25-14-13-20-21-16-18(28-2)11-12-22(21)26-15-6-9-17-8-4-5-10-19(17)24(20)26/h4-5,8,10-12,16H,3,6-7,9,13-15H2,1-2H3,(H,25,27)

InChI key

SPIFQXPRSJKRAO-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Melatonin receptor antagonist.

Features and Benefits

This compound is featured on the Melatonin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service