Skip to Content
Merck
All Photos(2)

Documents

HS15983G

Slide mailer, 5 place

end opening

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123302
NACRES:
NB.35

material

polypropylene

feature

Holds 5 x slides 76x26 mm

packaging

package of 25 Slide mailers

manufacturer/tradename

Heathrow Scientific HS15983G

color

assorted colors, Red, Yellow, Blue, Purple, Orange

Looking for similar products? Visit Product Comparison Guide

General description

Contains 5 of each color.
Ideal for shipping, storing or staining slides. These polypropylene containers hold 5 standard slides (3 × 1 in. or 25 × 75 mm). Slotted channels inside the mailer keep slides safely separated. Both end-opening and side-opening versions close securely to keep slides safe. The side-opening mailer measures 81 × 19 × 31 mm; end-opening mailer is 43 × 24 × 88 mm.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service