Skip to Content
Merck
All Photos(1)

Key Documents

H1751

Sigma-Aldrich

Hyaluronic acid potassium salt from human umbilical cord

Highly polymerized

Synonym(s):

Poly(β-glucuronic acid-[1→3]-β-N-acetylglucosamine-[1→4]), alternating

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C14H20KNO11)n
CAS Number:
MDL number:
UNSPSC Code:
12352201

impurities

<2% Protein (Lowry)

intrinsic viscosity

~100(lit.)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

High molecular weight polymer composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine which forms the core of complex proteoglycan aggregates found in extracellular matrix.

Quality

Also contains chondroitin sulfate.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dilara Pirhan et al.
Current eye research, 41(1), 59-69 (2015-02-07)
To evaluate the effects of the neuroprotective agents riluzole and resveratrol on the survival of retinal ganglion cells (RGCs) when administered alone or in combination. Experimental glaucoma was induced by injecting hyaluronic acid into the anterior chamber of Wistar albino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service