Skip to Content
Merck
All Photos(2)

Documents

GHS3128

Sigma-Aldrich

Hematoxylin Solution, Gill No. 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

6 g/L

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Gill formulation No. 3 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.. Used with hematoxylin and eosin staining.

Other Notes

6 g/L certified hematoxylin

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service