EP5702745006
Eppendorf® Adapter for 85/100 mL Round Bucket
pack of 2 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
Holds 1.5 - 2 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702745006
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL round buckets
Looking for similar products? Visit Product Comparison Guide
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service