Skip to Content
Merck
All Photos(1)

Key Documents

EP0030700112

Eppendorf® Cell Culture Dishes

Tissue Culture (TC)-treated surface, size 35 mm, case of 300 ea (Internal packs of 10)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.13

material

polystyrene

sterility

sterile

feature

lid

packaging

case of 300 ea (Internal packs of 10)

manufacturer/tradename

Eppendorf® 0030700112

diam. × H

36.7 mm × 10.8 mm

dish capacity

8.1 mL

dish diam.

36.7 mm

maximum volume

8.1 mL

size

35 mm

surface area

9.5 cm2

working volume

2.0-3.0 mL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Whenever you need direct access to your cells, dishes are the format of choice. Eppendorf cell culture dishes were designed with a focus on improved handling and stacking performance to ensure a new level of safe and carefree usage. It all starts with the packaging: the innovative design can be easily opened without tools and resealed without tape. The bags are shrinkable for space-saving, safe, and contamination-free storage of unused dishes. With the Eppendorf dishes, a corrugated handling ring provides unsurpassed safety in handling during transportation and expansion of cells. A SplashProtect ring inside the dish lid prevents spillage and traps fluid if condensation occurs. Combined with the outstanding, sturdy stacking, our cell culture dishes are unparalleled in their ease of transport and handling, safety, and convenience.

Innovative packaging
The packaging combines tool-free opening, resealable top and side for tape-free closing and an easy solution to shrink the package for space-conscious storage

Safety in all situations
A corrugated handling ring provides unsurpassed safety in handling during transportation and while working with dishes. The SplashProtect ring inside the dish lid traps liquid and prevents spills during transportation or incubation.

Direct surface identification
Easy surface reference and robust stacking performance by pronounced rims on dish lid and exact fitting of lid and bottom

Application

Expansion and culture of adherent and suspension cells on TC treated or on non-treated polystyrene

Performance of cell-based assays and microscopic analysis

Features and Benefits

Unsurpassed safety in handling during transportation and while working due to corrugated handling ring

SplashProtect ring inside of the dish lid traps liquid and prevents spills during transportation or incubation

Pronounced rim on dish lid and exact fitting of lid and bottom ensure sturdy, secure stacking

Easy differentiation of lid and dish bottom prevents unintended removal of the lid

Packaging combines tool-free opening, resealable top and side for tape-free closing, and an easy solution to shrink the package for space-conscious storage

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service