Recommended Products
product name
REM 885, 06070401
biological source
human lymph
growth mode
Suspension
karyotype
46,XY.ish dup(21)(q21)(RP11-15D13++,RP11-66H5++)
morphology
Lymphoblastoid
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Related Categories
Cell Line Origin
Human EBV transformed lymphocytes Alzheimer
Cell Line Description
Derived from a patient with early onset Alzheimer disease and cerebral amyloid angiopathy (CAA), carrying a 6.5 Mb microduplication of the APP gene locus on chromosome 21. This is an EBV-transformed lymphoblastoid cell line.
Culture Medium
RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 300,000 to 900,000 cells/ml; 5% CO2; 37°C
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service