Skip to Content
Merck
All Photos(1)

Key Documents

C2258

Sigma-Aldrich

Cellulose phosphate

(Fine Mesh)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817

Quality Level

form

(Fine Mesh)

capacity

~1.2 meq/g

Looking for similar products? Visit Product Comparison Guide

Application

Cellulose phosphate is used in protein chromatography, ion exchange chromatography and cation exchange media. Cellulose phosphate (CELLPHOS) has been studied as a collector for analytical preconcentration of traces of Cd(II), Cr(III), Cu(II) and Ni(II) from aqueous sample solution and used to purify and characterize acid phosphatase from approximately 3000-year-old human bones from archeological excavations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A N Carter
Current protocols in molecular biology, Chapter 18, Unit 18-Unit 18 (2008-02-12)
In studies of the regulation of specific biochemical events by reversible phosphorylation, assaying the protein kinases themselves can often lead to significant progress in understanding the mechanistic details of a system under study. This unit describes assays for a variety
Yaşar Demir et al.
Preparative biochemistry & biotechnology, 33(4), 311-320 (2003-11-11)
In this research, acid phosphatase was purified and characterized from approximately 3000-year-old human bones from archeological excavations. Using anion exchange chromatography, two isoenzymes, TrACP and TsACP, were isolated from the bone. TrACP and TsACP were eluted separately, with a concentration
N V Tsymbal et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 66(1), 29-35 (2004-04-24)
Nonspecific nuclease has been isolated from the cells of cyanobacterium Plectonema boryanum and purified to homogenic state. It has been established that the method of centrifugation of cell-free culture extract in the sucrose density gradient is efficient for the separation
Overexpression and purification of Saccharomyces cerevisiae DNA topoisomerase II from yeast.
J E Lindsley
Methods in molecular biology (Clifton, N.J.), 94, 187-197 (2003-07-08)
Assay of intrinsic transcript termination by E. coli RNA polymerase on single-stranded and double-stranded DNA templates.
Susan M Uptain
Methods in enzymology, 371, 339-351 (2004-01-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service