Skip to Content
Merck
All Photos(1)

Documents

B1893

Sigma-Aldrich

8-Bromoguanosine

Synonym(s):

2-Amino-8-bromo-6-hydroxypurine riboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12BrN5O5
CAS Number:
Molecular Weight:
362.14
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥99% (TLC)

form

solid

storage temp.

−20°C

SMILES string

NC1=Nc2c(nc(Br)n2[C@@H]3O[C@@H](CO)[C@H](O)[C@@H]3O)C(=O)N1

InChI

1S/C10H12BrN5O5/c11-9-13-3-6(14-10(12)15-7(3)20)16(9)8-5(19)4(18)2(1-17)21-8/h2,4-5,8,17-19H,1H2,(H3,12,14,15,20)/t2?,4?,5?,8-/m1/s1

InChI key

ASUCSHXLTWZYBA-RMAPPFHFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carla B Mellough et al.
Stem cells translational medicine, 8(7), 694-706 (2019-03-28)
A major goal in the stem cell field is to generate tissues that can be utilized as a universal tool for in vitro models of development and disease, drug development, or as a resource for patients suffering from disease or
David J Proctor et al.
Journal of the American Chemical Society, 125(9), 2390-2391 (2003-02-27)
In an effort to reduce the conformational heterogeneity of RNA, the modified nucleobase 8-bromoguanosine (8BrG) was introduced into oligonucleotides having the hairpin tetraloop motif YNMG (Y = U or C and M = C or A). Purine nucleobases with bromine
Elizabeth C Western et al.
The Journal of organic chemistry, 68(17), 6767-6774 (2003-08-16)
Modification of nucleosides to give pharmaceutically active compounds, mutagenesis models, and oligonucleotide structural probes continues to be of great interest. The aqueous-phase modification of unprotected halonucleosides is reported herein. Using a catalyst derived from tris(3-sulfonatophenyl)phosphine (TPPTS) and palladium acetate, 8-bromo-2'-deoxyguanosine
Leishmania amazonensis infection is reduced in macrophages treated with guanine ribonucleosides.
S Giorgio et al.
Acta tropica, 70(1), 119-122 (1998-08-26)
Julia Dolinina et al.
American journal of physiology. Renal physiology, 311(5), F984-F990 (2016-09-30)
There is increasing evidence that the permeability of the glomerular filtration barrier (GFB) is partly regulated by a balance between the bioavailability of nitric oxide (NO) and that of reactive oxygen species (ROS). It has been postulated that normal or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service