Skip to Content
Merck
All Photos(1)

Key Documents

AV46013

Sigma-Aldrich

Anti-PRIM1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-MGC12308, Anti-Primase, polypeptide 1, 49 kDa, Anti-p49

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

50 kDa

species reactivity

rabbit, rat, guinea pig, bovine, human, horse, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRIM1(5557)

Immunogen

Synthetic peptide directed towards the N terminal region of human PRIM1

Application

Anti-PRIM1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml.

Biochem/physiol Actions

Human DNA primase 1 (PRIM1) is a pivotal enzyme component of complex chromosomal replication apparatus. It functions in synchronization with DNA polymerase alpha during replication in eukaryotic cells. DNA primase facilitates the synthesis of small RNA primers that initiate synthesis of the lagging DNA strand.

Sequence

Synthetic peptide located within the following region: SQYYRWLNYGGVIKNYFQHREFSFTLKDDIYIRYQSFNNQSDLEKEMQKM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Stadlbauer et al.
European journal of biochemistry, 222(3), 781-793 (1994-06-15)
DNA-polymerase-alpha--primase complex contains four subunits, p180, p68, p58, and p48, and comprises a minimum of two enzymic functions. We have cloned cDNAs encoding subunits of DNA-polymerase-alpha--primase from human and mouse. Sequence comparisons showed high amino acid conservation among the mammalian
Mairi L Kilkenny et al.
The Journal of biological chemistry, 287(28), 23740-23747 (2012-05-18)
The DNA polymerase α-primase complex forms an essential part of the eukaryotic replisome. The catalytic subunits of primase and pol α synthesize composite RNA-DNA primers that initiate the leading and lagging DNA strands at replication forks. The physical basis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service