Skip to Content

A8731

Anti-Atg12 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-APG12, Anti-Autophagy related 12 homolog (S. cerevisiae)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
14
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~20 kDa

species reactivity

mouse

technique(s)

immunoprecipitation (IP): 5-10 μL using whole extract of HEK-293T cells co-expressing mouse Atg12 and Atg5-EGFP proteins, western blot: 1:250-1:500 using whole extract of HEK-293T cells expressing mouse Atg12

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATG12(9140)
mouse ... Atg12(67526)

General description

Atg12 is an autophagy-related protein. It forms a complex with Atg5 and functions in LC3 processing.
The autophagy-related protein 3 (ATG3) gene is mapped to human chromosome 5q22.3. ATG12 is a mammalian homolog of yeast Apg3p/ Atg3 is ubiquitously expressed in human tissues.

Immunogen

synthetic peptide corresponding to amino acids 27-39 of mouse Atg12, conjugated to KLH. The corresponding sequence differs by one amino acid in human.

Application

Anti-Atg12 antibody produced in rabbit has been used for:
  • immunoprecipitation
  • western blot
  • immunohistochemistry staining.

Biochem/physiol Actions

Macroautophagy, usually referred to as autophagy, is a major pathway for bulk degradation of cytoplasmic constituents and organelles. Atg12 is a ubiquitin-like modifier essential for autophagy. Two ubiquitin-like conjugation systems are involved in autophagosome formation: Atg12 and Atg8 conjugation systems. Autophagy is required for normal turnover of cellular components during starvation. It plays an essential role in cellular differentiation, cell death and aging. Defective autophagy may contribute to certain human diseases such as cancer, neurodegenerative diseases, muscular disorders and pathogen infections.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library