A8255
5α-Androstane-3,17-dione
≥99%
Synonym(s):
Androstanedione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
≥99%
drug control
USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada
SMILES string
C[C@]12CCC(=O)C[C@@H]1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CCC4=O
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
199.9 °F - closed cup
Flash Point(C)
93.3 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Julia V Bukanova et al.
Frontiers in molecular neuroscience, 13, 44-44 (2020-04-09)
The ability of androstane and androstene neurosteroids with modifications at C-17, C-5, and C-3 (compounds 1-9) to influence the functional activity of inhibitory glycine and γ-aminobutyric acid (GABA) receptors was estimated. The glycine- and GABA-induced chloride current (IGly and IGABA)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service