Skip to Content
Merck
All Photos(1)

Key Documents

A8170

Sigma-Aldrich

Ammonium citrate dibasic

ACS reagent, 98.0-103.0%

Synonym(s):

Ammonium hydrogencitrate, Citric acid ammonium salt, Diammonium hydrogen citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(CO2H)(CH2CO2NH4)2
CAS Number:
Molecular Weight:
226.18
Beilstein:
4925760
EC Number:
MDL number:
UNSPSC Code:
12161700

grade

ACS reagent

vapor density

1.8 (vs air)

Assay

98.0-103.0%

impurities

≤0.005% Insoluble matter

ign. residue

≤0.01%

anion traces

chloride (Cl-): ≤0.001%
oxalate (C2O42-): passes test (limit about 0.005%)
phosphate (PO43-): ≤5 ppm
sulfur compounds (as SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤5 ppm

SMILES string

N.N.OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7.2H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);2*1H3

InChI key

YXVFQADLFFNVDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service