Product Name
Atto 680, BioReagent, suitable for fluorescence, ≥90% (HPLC)
InChI
1S/C27H31N3O6S/c1-4-29-9-5-7-17-11-20-24(13-22(17)29)36-25-14-23-19(12-21(25)28-20)18(16-37(33,34)35)15-27(2,3)30(23)10-6-8-26(31)32/h11-15H,4-10,16H2,1-3H3,(H-,31,32,33,34,35)
InChI key
MHHMNDJIDRZZNT-UHFFFAOYSA-N
product line
BioReagent
assay
≥90% (HPLC)
form
powder
manufacturer/tradename
ATTO-TEC GmbH
fluorescence
λex 680 nm; λem 695 nm in 0.1 M phosphate pH 7.0
suitability
suitable for fluorescence
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Atto 680 shows a molar extinction of 120,000 and QY of 30% in water (40% in ethanol). Decay time is 1.8 ns.
General description
Other Notes
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service