Skip to Content
Merck
All Photos(1)

Key Documents

6619

Phosphorus Standard

Synonym(s):

Potassium Phosphate Monobasic solution, Monopotassium phosphate, Potassium dihydrogen phosphate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51000000

IVD

for in vitro diagnostic use

storage temp.

2-8°C

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Phosphorus (as KH2PO4), 20 ug/mL, in 0.05 N HCl. Equivalent to 645 nmol P/mL.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service