Skip to Content
Merck
All Photos(1)

Documents

131-500A

Sigma-Aldrich

Keratinocyte Serum-Free Growth Medium for adult cells (500 ml)

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media

Application

Keratinocyte Serum-Free Growth Medium for adult cells (500 ml) has been used in the culturing of human bronchial epithelial cells.

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Werner E G Müller et al.
Cancers, 13(4) (2021-03-07)
The anti-cancer antitumor antibiotic bleomycin(s) (BLM) induces athyminic sites in DNA after its activation, a process that results in strand splitting. Here, using A549 human lung cells or BEAS-2B cells lunc cells, we show that the cell toxicity of BLM

Protocols

Storage - Preparation for Culturing - Culturing HEK - Subculturing HEK

Storage - Preparation for Culturing - Culturing HEK - Subculturing HEK

Storage - Preparation for Culturing - Culturing HEK - Subculturing HEK

Storage - Preparation for Culturing - Culturing HEK - Subculturing HEK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service