Recommended Products
Assay
≥98.0% (T)
shelf life
limited shelf life
pKa
3.1
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Other Notes
Immobilized pH 2.5-11 gradients for 2-D electrophoresis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P Sinha et al.
Electrophoresis, 13(4), 210-214 (1992-04-01)
Extremely wide immobilized pH gradients, pH 2.5-11, for isoelectric separation of complex protein mixtures are described. These pH gradients are theoretically and practically the maximum that can be achieved at present with the available acrylamido buffers and titrants. Conditions are
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service