Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C17H18N2O
CAS Number:
Molecular Weight:
266.34
UNSPSC Code:
41116107
PubChem Substance ID:
MDL number:
grade
analytical standard
form
powder
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
color
tan
application(s)
forensics and toxicology
pharmaceutical (small molecule)
veterinary
format
neat
storage temp.
−20°C
SMILES string
NCCc1c[nH]c2c(OCc3ccccc3)cccc12
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.