Skip to Content
Merck

91077

tert-Butyl 2,2,2-trichloroacetimidate

purum, ≥97.0% (GC)

Synonym(s):

TBTA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CCl3C(=NH)OC(CH3)3
CAS Number:
Molecular Weight:
218.51
PubChem Substance ID:
UNSPSC Code:
12352100
Beilstein/REAXYS Number:
1770049
MDL number:
Assay:
≥97.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

Quality Level

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.456 (lit.), n20/D 1.457

bp

65 °C/11 mmHg (lit.)

mp

21 °C (lit.)

density

1.221 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=N)C(Cl)(Cl)Cl

InChI

1S/C6H10Cl3NO/c1-5(2,3)11-4(10)6(7,8)9/h10H,1-3H3

InChI key

CQXDYHPBXDZWBA-UHFFFAOYSA-N

Other Notes

Reagent used for the smooth and selective preparation of tert-butyl esters and ethers from acids and alcohols, respectively


Still not finding the right product?


pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

131.0 °F - closed cup

flash_point_c

55 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A. Armstrong et al.
Tetrahedron Letters, 29, 2483-2483 (1988)
Felipe de Jesus Cortez et al.
PloS one, 15(11), e0242049-e0242049 (2020-11-14)
Islet autoantibodies are predominantly measured by radioassay to facilitate risk assessment and diagnosis of type 1 diabetes. However, the reliance on radioactive components, large sample volumes and limited throughput renders radioassay testing costly and challenging. We developed a multiplex analysis
Zhimin Ou et al.
Cell reports, 26(11), 2984-2997 (2019-03-14)
The CNS plays a pivotal role in energy homeostasis, but whether oligodendrocytes are involved has been largely unexplored. Here, we show that signaling through GPR17, a G-protein-coupled receptor predominantly expressed in the oligodendrocyte lineage, regulates food intake by modulating hypothalamic neuronal



Global Trade Item Number

SKUGTIN
91077-25ML04061836828110
91077-5ML04061836828127