68570
3-Methyl-1-pentyn-3-ol
purum, ≥98.0% (GC)
Synonym(s):
Ethyl ethynyl methyl carbinol, Meparfynol
About This Item
vapor density
3 (vs air)
vapor pressure
6.5 mmHg ( 20 °C)
grade
purum
Assay
≥98.0% (GC)
refractive index
n20/D 1.431 (lit.)
n20/D 1.431
bp
121-122 °C (lit.)
density
0.866 g/mL at 25 °C (lit.)
SMILES string
CCC(C)(O)C#C
InChI
1S/C6H10O/c1-4-6(3,7)5-2/h1,7H,5H2,2-3H3
InChI key
QXLPXWSKPNOQLE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
82.4 °F - closed cup
Flash Point(C)
28 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service