63678
(−)-Menthone
technical, ≥90% (sum of enantiomers, GC)
Synonym(s):
(1R,4S)-p-Menthan-3-one, (2S,5R)-2-Isopropyl-5-methylcyclohexanone
About This Item
vapor pressure
0.5 mmHg ( 20 °C)
grade
technical
Assay
≥90% (sum of enantiomers, GC)
refractive index
n20/D 1.45 (lit.)
n20/D 1.451
bp
207-210 °C (lit.)
density
0.893 g/mL at 20 °C (lit.)
storage temp.
2-8°C
SMILES string
CC(C)[C@@H]1CC[C@@H](C)CC1=O
InChI
1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3/t8-,9+/m1/s1
InChI key
NFLGAXVYCFJBMK-BDAKNGLRSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
165.2 °F
Flash Point(C)
74 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service