Skip to Content
Merck
All Photos(1)

Documents

45986

Supelco

3-Nitrotoluene

analytical standard

Synonym(s):

1-Methyl-3-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NO2
CAS Number:
Molecular Weight:
137.14
Beilstein:
1906910
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.73 (vs air)

vapor pressure

1 mmHg ( 50.2 °C)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.541 (lit.)

bp

230-231 °C (lit.)

mp

14-16 °C (lit.)

density

1.157 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1cccc(c1)[N+]([O-])=O

InChI

1S/C7H7NO2/c1-6-3-2-4-7(5-6)8(9)10/h2-5H,1H3

InChI key

QZYHIOPPLUPUJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Nitrotoluene is an organic compound belonging to the class of nitrobenzenes. It is encountered in forensic science and is widely used as raw material and intermediaries in dyes, rubber chemicals, pharmaceuticals and pesticides.

Application

3-Nitrotoluene may be used as an analytical reference standard for the quantification of the analyte in post-blast debris samples and water samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

208.4 °F

Flash Point(C)

98 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasound-assisted magnetic solid-phase extraction based ionic liquid-coated Fe 3 O 4@ graphene for the determination of nitrobenzene compounds in environmental water samples
Cao X, et al.
Analyst, 139(8), 1938-1944 (2014)
Highly sensitive screening method for nitroaromatic, nitramine and nitrate ester explosives by high performance liquid chromatography?atmospheric pressure ionization-mass spectrometry (HPLC-API-MS) in forensic applications
Xu X, et al.
Journal of Forensic Sciences, 49(6) (2004)
G Allenby et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 16(4), 710-724 (1991-05-01)
A method for culturing isolated seminiferous tubules (ST) from adult rats for 1-3 days has been developed and optimized rigorously on the basis of the secretion of immunoactive inhibin under basal conditions and after maximal stimulation with rat FSH or
Muhammad Shariq Vohra et al.
Water research, 36(1), 59-64 (2002-01-05)
TiO2-mediated photocatalytic degradation process was employed to treat aqueous 2-, 3- and 4-NT (nitrotoluene) pollutants. The NT disappearance and TOC removal rates for three isomers showed no significant differences. Three hydroxylated aromatic intermediates resulting from the photocatalytic degradation of 4-NT
J P Chism et al.
Drug metabolism and disposition: the biological fate of chemicals, 13(6), 651-657 (1985-11-01)
2-Nitrotoluene (2NT) induces DNA repair in the in vivo-in vitro hepatocyte unscheduled DNA synthesis assay in male, but not female, Fischer-344 rats. 3-Nitrotoluene (3NT) and 4-nitrotoluene (4NT) are inactive in both sexes. The structurally related rat hepatocarcinogen, 2,6-dinitrotoluene, which also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service