Skip to Content
Merck
All Photos(1)

Key Documents

44880

Supelco

1-Phenyleicosane

analytical standard

Synonym(s):

Eicosylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)19C6H5
CAS Number:
Molecular Weight:
358.64
Beilstein:
2561141
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

40-43 °C

application(s)

environmental

SMILES string

CCCCCCCCCCCCCCCCCCCCc1ccccc1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service