Skip to Content
Merck
All Photos(1)

Documents

18309

Supelco

Ginkgolide C

analytical standard

Synonym(s):

1,7-Dihydroxy-ginkgolide A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O11
CAS Number:
Molecular Weight:
440.40
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

[H][C@@]12OC(=O)[C@@H](C)[C@]1(O)[C@@]34OC5OC(=O)[C@H](O)C56[C@@H]([C@H](O)[C@@H](OC3=O)[C@@]46[C@H]2O)C(C)(C)C

InChI

1S/C20H24O11/c1-5-12(24)28-11-8(22)18-10-6(21)7(16(2,3)4)17(18)9(23)13(25)30-15(17)31-20(18,14(26)29-10)19(5,11)27/h5-11,15,21-23,27H,1-4H3/t5-,6+,7+,8+,9+,10-,11+,15?,17?,18-,19-,20-/m1/s1

InChI key

AMOGMTLMADGEOQ-CFTQXCTRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginkgolide C is an active constituent of Ginkgo biloba. The ginkgo leaves has several pharmacological properties, some of them being; improved blood flow, radical scavenging, vasoprotection, and anti-PAF (platelet aggregating factor) activity. Ginkgolide C along with the other active constituents of Ginkgo biloba, is responsible for the inhibition of PAF activity.

Application

It may be used as a reference standard in determination of ginkgolide from Ginkgo biloba commercial product, using HPLC with evaporative light-scattering detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: ginkgo

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative analysis of bilobalide and ginkgolides from Ginkgo biloba leaves and Ginkgo products using (1)H-NMR.
Choi YH
Chemical & Pharmaceutical Bulletin, 51(2), 158-161 (2003)
Rapid determination of terpene lactones in Ginkgo Biloba commercial products by HPLC with evaporative light-scattering detection.
Herring, Tim.
LCGC North America, 22.5, 456-462 (2004)
Haixia Lin et al.
Toxicological sciences : an official journal of the Society of Toxicology, 139(2), 338-349 (2014-03-07)
Ginkgo biloba has been used for many thousand years as a traditional herbal remedy and its extract has been consumed for many decades as a dietary supplement. Ginkgo biloba leaf extract is a complex mixture with many constituents, including flavonol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service