Skip to Content
Merck
All Photos(1)

Documents

04313

Millipore

Fluorocult® DEV Lactose Peptone Broth

suitable for microbiology, NutriSelect® Basic, FluoroCult®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bromocresol purple, 0.02 g/L
lactose, 10.0 g/L
4-methylumbelliferyl-β-D-glucuronidase, 0.1 g/L
peptone from casein, 17.0 g/L
peptone from soya, 3.0 g/L
sodium chloride, 5.0 g/L
tryptophan, 1.0 g/L

manufacturer/tradename

FluoroCult®
NutriSelect® Basic

final pH

7.2±0.2 (25 °C)

application(s)

environmental
food and beverages
water monitoring

microbiology

suitability

Escherichia coli
coliforms

Related Categories

Application

This medium is particularly suited for the enrichment and determination of the titre of coliform bacteria in the bacteriological analysis of water. E. coli shows a positive fluorescence under UV light (366 nm). A positive indole reaction is made for confirmation.

Preparation Note

Suspend 36.1 g/litre or 72.2 g/litre, dispense into test tubes fitted with DURHAM tubes, autoclave (15 min at 121° C).

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

FLUOROCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service