Skip to Content
Merck
All Photos(1)

Documents

00700

Sigma-Aldrich

Acetonitrile

≥99.5% (GC), ACS reagent, puriss. p.a.

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

product name

Acetonitrile, puriss. p.a., ACS reagent, ≥99.5% (GC)

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

1.41 (vs air)
1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)
72.8 mmHg

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %
8 % (lit.)

impurities

≤0.001% free alkali (as NH3)
≤0.005% free acid (as CH3COOH)
≤0.1% benzene (GC)
≤0.2% propionitrile (GC)
≤0.2% water

evapn. residue

≤0.001%

refractive index

n20/D 1.344 (lit.)
n20/D 1.344

bp

81-82 °C (lit.)
81-82 °C (lit.)

mp

-48 °C (lit.)
−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.05 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CC#N

format

neat

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile (MeCN) is a polar solvent.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of hydroxypyrazoles and 1-methyl-3-isoxazolones via haloform reactions.
Flores AFC, et al.
Tetrahedron Letters, 43(28), 5005-5008 (2002)
Lavanya Premvardhan et al.
Biochimica et biophysica acta, 1847(10), 1187-1199 (2015-05-20)
The structures and environments of the protein-bound peridinins (Pers) and chlorophylls (Chls) a/c2 in the membrane-intrinsic major light-harvesting complex of the dinoflagellate Amphidinium carterae (LHCAmph) are characterised using resonance Raman (RR) spectroscopy with 11 excitation wavelengths, at 77K. The excitation-dependent
Michelangelo Anastassiades et al.
Journal of AOAC International, 86(2), 412-431 (2003-05-02)
A simple, fast, and inexpensive method for the determination of pesticide residues in fruits and vegetables is introduced. The procedure involves initial single-phase extraction of 10 g sample with 10 mL acetonitrile, followed by liquid-liquid partitioning formed by addition of
Timo Glatter et al.
Molecular systems biology, 5, 237-237 (2009-01-22)
Protein complexes represent major functional units for the execution of biological processes. Systematic affinity purification coupled with mass spectrometry (AP-MS) yielded a wealth of information on the compendium of protein complexes expressed in Saccharomyces cerevisiae. However, global AP-MS analysis of
Elke H J Krekels et al.
Clinical pharmacokinetics, 53(6), 553-563 (2014-02-06)
From a previously validated paediatric population pharmacokinetic model, it was derived that non-linear morphine maintenance doses of 5 μg/kg(1.5)/h, with a 50 % dose reduction in neonates with a postnatal age (PNA) <10 days, yield similar morphine and metabolite concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service