Skip to Content
Merck
All Photos(1)

Key Documents

00166

Supelco

Eschka′s Reagent

for sulfur determination in carbon, 38-42% (Mg, complexometric)

Synonym(s):

Magnesium oxide – sodium carbonate 2:1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2MgO · Na2CO3
CAS Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

38-42% (Mg, complexometric)

form

powder

quality

for sulfur determination in carbon

impurities

magnesium oxide
≤0.0005% total of sulfur (S)

loss

≤0.5% loss on drying, 105 °C

cation traces

Ba: ≤10 mg/kg
Ca: ≤200 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

forensics and toxicology

SMILES string

[Na+].[Na+].O=[Mg].O=[Mg].[O-]C([O-])=O

InChI

1S/CH2O3.2Mg.2Na.2O/c2-1(3)4;;;;;;/h(H2,2,3,4);;;;;;/q;;;2*+1;;/p-2

InChI key

GANAIKRVCIWEMO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service