Skip to Content
Merck
All Photos(1)

Key Documents

YY2004076

Millipore

Adapter 1-1/2 in. TC to 14 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161525
eCl@ss:
32031603

I.D.

3.8 cm (1.5 in.)

Quality Level

General description

The Adapter 1-1/2 in. TC to 14 mm is intended for use with the 293 mm stainless steel sanitary filter holder - product number YY3029316 - an ideal solutions for sterilizing liquids or gases by pressure filtration.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service