Skip to Content
Merck
All Photos(1)

Key Documents

ST1684

Sigma-Aldrich

Anti-MAG Mouse mAb (3C7)

liquid, clone 3C7, Calbiochem®

Synonym(s):

Anti-Myelin Associated Glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3C7, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAG(4099)

General description

Purified mouse monoclonal antibody. Recognizes the ~50-52 kDa MAG protein.
Recognizes the ~50-52 kDa MAG protein in Jurkat cells and human cerebellum tissue.
This Anti-MAG Mouse mAb (3C7) is validated for use in Immunoblotting Paraffin Sections for the detection of MAG.

Immunogen

A recombinant polypeptide corresponding to amino acids of 119-209 human MAG, expressed as a GST fusion protein

Application

Immunoblotting (1-5 µg/ml)Paraffin Sections (3 µg/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Jurkat cells, human cerebellum tissue

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolin Hou et al.
Molecular medicine reports, 17(3), 4515-4523 (2018-01-19)
Oligodendrocytes (OLs) are myelin-forming cells that are present within the central nervous system. Impaired oligodendrocyte precursor cell (OPC) differentiation into mature OLs is a major cause of demyelination diseases. Therefore, identifying the underlying molecular mechanisms of OPC differentiation is crucial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service