MABN877
Anti-NB-2/Cntn5 Antibody, clone 1A6
clone 1A6, from rat
Synonym(s):
Contactin-5, Neural recognition molecule NB-2
About This Item
Recommended Products
biological source
rat
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
1A6, monoclonal
species reactivity
mouse
technique(s)
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
mouse ... Cntn5(244682)
General description
Specificity
Immunogen
Application
Neuroscience
Developmental Neuroscience
Immunocytochemistry Analysis: A representative lot detected punctate NB-2 immunoreactivity on the dendrites of cultured embryonic E17 mouse hippocampal neurons. A partial overlap of NB-2 immunoreactivity with those of APLP1 and synapsin was seen (Shimoda, Y., et al. (2012). Neurosci. Lett. 510(2):148-153).
Immunofluorescence Analysis: A representative lot detected NB-2 immunoreactivity among parvalbumin-/PV-positive proprioceptive sensory neurons by fluorescent immunohistochemistry staining of postnatal day 6 (p6) mouse dorsal root ganglion (DRG) cryosectoins (Ashrafi, S., et al. (2014). Neuron. 81(1):120-129).
Western Blotting Analysis: A representative lot detected NB-2 in presynaptic preparations from the spinal cords of postnatal p6 to p7 mice (Ashrafi, S., et al. (2014). Neuron. 81(1):120-129).
Western Blotting Analysis: A representative lot detected NB-2 in the synaptosomal fractions from mouse cerebral cortex homogenate (Shimoda, Y., et al. (2012). Neurosci. Lett. 510(2):148-153).
Quality
Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be IgG2a.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service