MABF723
Anti-CD86 (B7-2) Antibody (mouse), APC, clone GL-1
clone GL-1 (GL1), 0.2 mg/mL, from rat
Synonym(s):
T-lymphocyte activation antigen CD86, Activation B7-2 antigen, Early T-cell costimulatory molecule 1, ETC-1, CD86
About This Item
Recommended Products
biological source
rat
Quality Level
conjugate
Allophycocyanin conjugate
antibody form
purified antibody
antibody product type
primary antibodies
clone
GL-1 (GL1), monoclonal
species reactivity
mouse
packaging
antibody small pack of 25 μg
concentration
0.2 mg/mL
technique(s)
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
isotype
IgG2aκ
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Cd86(12524)
Related Categories
General description
Immunogen
Application
Inflammation & Immunology
Quality
Flow Cytometry Analysis: 0.06 μg of this antibody detected CD86 (B7-2) in one million C57Bl/6 splenocytes.
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service