Skip to Content
Merck
All Photos(1)

Key Documents

MABE949

Sigma-Aldrich

Anti-E3 Ubiquitin Ligase BRE1B Antibody, clone 4H8.1

clone 4H8.1, from mouse

Synonym(s):

E3 ubiquitin-protein ligase BRE1B, 95 kDa retinoblastoma-associated protein, BRE1-B, RBP95, RING finger protein 40, Syntaxin-1-interacting RING finger protein, Protein staring

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H8.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... RNF40(9810)

General description

E3 ubiquitin-protein ligase BRE1B (UniProt O75150; also known as 95 kDa retinoblastoma-associated protein, BRE1-B, RBP95, RING finger protein 40, Syntaxin-1-interacting RING finger protein, Protein staring) is encoded by the RNF40 (also known as BRE1B, KIAA0661) gene (Gene ID 9810) in human. Proteini ubiquitination is mediated by ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin ligases (E3s). There exist RING finger-type (really interesting new gene), HECT-type (homologous to E6-AP C-terminus), and U-box type of E3 ligases. RING finger ubiquitin ligase members can function as monomers, dimers, or multi-subunit complexes. The RNF20-RNF40 heterodimer mediates the ubiquitination of histone H2B at Lysine 120, an event that plays an important role in decompacting chromatin fiber to allow transcription elongation, in stem cell differentiation, as well as in DNA double-strand breaks (DSBs) repair via the homologous recombination (HR) and nonhomologous end-joining (NHEJ) pathways. Human RNF40 is a 1001-amino acid protein containing a RING-type zinc finger in its C-terminal region (a.a. 948-987) that mediates its interaction with RNF20.

Specificity

Clone 4H8.1 epitope region is present in all three spliced isoforms of human BRE1B reported by UniProt (O75150).

Immunogen

Epitope: Internal (C-terminal half).
GST-tagged recombinant internal fragment from the C-terminal half of human BRE1B .

Application

Anti-E3 Ubiquitin Ligase BRE1B Antibody, clone 4H8.1 is an antibody against E3 Ubiquitin Ligase BRE1B for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected BRE1B in 10 µg of HeLa cell lysate.

Target description

~120 kDa oberved. 113.7 kDa (isoform 1), 94.80 kDa (isoform 2), 102.0 kDa (isoform 3) calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service