Skip to Content
Merck

MABC681

Anti-ETV4 Antibody, clone 1A2G3

ascites fluid, clone 1A2G3, from mouse

Synonym(s):

ETS translocation variant 4, Adenovirus E1A enhancer-binding protein, E1A-F, Polyomavirus enhancer activator 3 homolog, Protein PEA3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1A2G3, monoclonal
Application:
WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1A2G3, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ETV4(2118)

General description

ETS translocation variant 4 protein, alternatively known as Adenovirus E1A enhancer-binding protein E1A-F, or Polyomavirus enhancer activator 3 homolog (PEA3), is encoded by the human gene named ETVA/E1AF/PEA3. ETV4 is a protein that is a transcriptional activator that was first recognized as such because it allowed for the expression of the adenovirus E1A gene upon infection. Today ETV4 has been shown to be active in a wide variety of biological processes from stem cell differentiation, to mammary gland and duct morphogenesis to motor neuron and axon guidance. ETV4 activity is regulated via the ERK/MAP signaling and a complex interplay of post-translation modifications such as sumoylation and ubiquitination. In cancers, ETV4 activates HIF and its subsequent proliferation genes and overexpression in prostate cells leads to oncogenesis. EMD-Millipore’s Anti-ETV4 mouse monoclonal antibody has been tested in western blot against K562 cell lysates as well as recombinantly expressed ETV4 protein.
~54 kDa observed. Uncharacterized bands may appear in some lysate(s).

Immunogen

Purified recombinant fragment of human ETV4 expressed in E. Coli.

Application

Detect ETS translocation variant 4 protein using this mouse monoclonal antibody, Anti-ETV4 Antibody & clone 1A2G3 validated for use in western blotting.

Analysis Note

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected ETV4 in K562 cell lysate.

Optimal working dilutions must be determined by end user.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library