Skip to Content
Merck
All Photos(3)

Documents

MABC663

Sigma-Aldrich

Anti-TICAM1 Antibody, clone 3D5.1

clone 3D5.1, from mouse

Synonym(s):

TICAM1, TIR domain-containing adapter molecule 1, TICAM-1, Proline-rich, vinculin and TIR domain-containing protein B, Putative NF-kappa-B-activating protein 502H, Toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta, My

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D5.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TICAM1(148022)

General description

TIR domain-containing adapter molecule 1 (UniProt Q8IUC6), also known as MyD88-3, Proline-rich, vinculin and TIR domain-containing protein B, Putative NF-kB-activating protein 502H, TICAM-1, TIR domain containing adaptor inducing interferon-b, TIR domain-containing adapter protein inducing IFN-b, Toll-interleukin-1 receptor domain-containing adapter protein inducing interferon b) is encoded by the TICAM1 gene in human. TCAM-1/TRIF/MyD88-3 is as an adaptor molecule that mediates Toll-like receptor 3 (TLR3)-dependent NF-kB activation and cytokine expression. Upon recognition of double-stranded RNA, TLR3 recruits TRIF, which in turn associates with TNF receptor-associated factor 3 (TRAF3) to activate two Ik-B kinase (IKK)-related kinases, TANK-binding kinase 1 (TBK1) and IKKi. These two kinases phosphorylate interferon regulatory factor 3/7 (IRF3/7), resulting in the induction of type I interferons (IFNs) and expression of IFN-inducible genes. TICAM-1 can also mediate NF-kB activation through receptor-interacting protein 1 (RIP1) and TRAF6. Human enterovirus 68 (EV68) is reported to disable TLR3 pathway signaling by cleaving TRIF in infected cells.

Immunogen

GST-tagged recombinant protein corresponding to human TICAM1.

Application

Anti-TICAM1 Antibody, clone 3D5.1 is an antibody against TICAM1 for use in Western Blotting, Immunohistochemistry, Immunocytochemistry.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected TICAM1 in human liver tissue.
Immunocytochemistry Analysis: 20 µg/mL from a representative lot detected TICAM1 in HeLa and A431 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected TICAM1 in 10 µg of Raji cell lysate.

Target description

~76 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service