Skip to Content
Merck
All Photos(2)

Documents

ABT337

Sigma-Aldrich

Anti-NANOS1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nanos homolog 1, NOS1, NANOS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NANOS1(340719)

General description

Nanos homolog 1 (NANOS1, NOS-1) contains one nanos-type zinc finger and belongs to the nanos family. It is an RNA-binding protein that is predominantly expressed in testis during germline development. NANOS 1 is highly expressed in lung carcinomas that can be detected in invasive tumor cells. NANOS1 up-regulates the production of MMP14 to promote tumor cell invasion.

Specificity

This antibody recognizes the intermediate domain of NANOS1.

Immunogen

Epitope: Intermediate domain
KLH-conjugated linear peptide corresponding to the intermediate domain of human NANOS1.

Application

Detect Nanos homolog 1 using this rabbit polyclonal antibody, Anti-NANOS1 Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: 2.5 µg/mL from a representative lot detected NANOS1 in human brain tissue.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected NANOS1 in 15 µg of rat brain tissue lysate.

Target description

~42 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service