Skip to Content
Merck
All Photos(1)

Key Documents

AB16311

Sigma-Aldrich

Anti-Nitric Oxide Synthase II Antibody

Chemicon®, from rabbit

Synonym(s):

NOS II, iNOS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reactive with mouse NOS-II (iNOS).

Immunogen

Synthetic peptide corresponding to amino acids 1131-1144 of mouse macrophage NOS, coupled to KLH.

Application

This Anti-Nitric Oxide Synthase II Antibody is validated for use in WB, IC for the detection of Nitric Oxide Synthase II.
Western Blot: 5 μg/mL using chemiluminescent detection. Higher concentrations of the antibody may be required if colorimetric detection is used. Antibody dilution will also vary with the concentration of the antigen. Immunocytochemistry: Use at 2-5 μg/mL

Optimal working dilutions must be determined by end user.

Linkage

Replaces: AB1631

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatriz Linillos-Pradillo et al.
International journal of molecular sciences, 24(18) (2023-09-28)
The liver is the organ responsible for the metabolism and detoxification of BPF, the BPA analogue that is replacing it in plastic-based products. It is not known whether BPF can trigger inflammatory responses via the NLRP3 inflammasome, which plays a
Bleranda Zeka et al.
Acta neuropathologica communications, 4(1), 82-82 (2016-08-10)
Neuromyelitis optica/spectrum disorder (NMO/SD) is a severe, inflammatory disease of the central nervous system (CNS). In the majority of patients, it is associated with the presence of pathogenic serum autoantibodies (the so-called NMO-IgGs) directed against the water channel aquaporin 4
Jordon Dunham et al.
Journal of neuropathology and experimental neurology, 76(6), 467-478 (2017-05-16)
Oxidative damage and iron redistribution are associated with the pathogenesis and progression of multiple sclerosis (MS), but these aspects are not entirely replicated in rodent experimental autoimmune encephalomyelitis (EAE) models. Here, we report that oxidative burst and injury as well
Isabella Wimmer et al.
Acta neuropathologica communications, 7(1), 14-14 (2019-02-02)
Human inflammatory or neurodegenerative diseases, such as progressive multiple sclerosis (MS), occur on a background of age-related microglia activation and iron accumulation as well as pre-existing neurodegeneration. Most experimental models for CNS diseases, however, are induced in rodents, which are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service