920078
Safety carrier
made to hold 2.5 L glass bottles
Synonym(s):
Transport carrier for 1l and 2.5 l bottles
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
compatibility
made to hold 2.5 L glass bottles
shipped in
ambient
General description
This specially developed safety carrier provides an increased protection against glass breakage. The solvent-resistant materials and the high compression strength of the inlay maximize the protection of 4-L-glass-bottles from us during transportation. The high inner volume (20 l) and the leak proof top cover protect against the contact with solvents and vapours in case of glass breakage.
for2.5 L glass bottles
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service