Skip to Content
Merck
All Photos(1)

Key Documents

6.17578

Supelco

di-Ammonium hydrogen phosphate

purified grade

Synonym(s):

di-Ammonium hydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
MDL number:
UNSPSC Code:
12352302
grade:
purified grade

grade

purified grade

SMILES string

[P](=O)([O-])([O-])[O-].[N+H4].[N+H4].[H+]

InChI

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service