Skip to Content
Merck

539746

InhibitorSelect 96-Well Protein Kinase Inhibitor Library III - Calbiochem

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 1 96-well plate(s)

General description

The InhibitorSelect 96-Well Protein Kinase Inhibitor Library III consists of 84, well-characterized protein kinase inhibitors, the majority of which are cell-permeable and ATP-competitive. This serine/threonine-selective library will be useful for target identification in drug discovery, biochemical pathway analysis, screening new protein kinases, and other pharmaceutical-related applications.

Physical form

81 inhibitors are supplied at a concentration of 10 mM and 3 inhibitors are supplied at a concentration of 5 mM in DMSO (50 μl/well).

Preparation Note

The contents of this pouch have been purged under nitrogen and are vacuum sealed. Cut open the pouch, take the plate out of the pouch, and let it thaw at room temperature. Please remove the lid gently. To re-store, purge with an inert gas, replace the lid

Other Notes

Supplied with a CD containing comprehensive documentation for each inhibitor. Please contact technical service for further details.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library