Skip to Content
Merck
All Photos(1)

Documents

200241

Sigma-Aldrich

6-Benzylaminopurine

Plant cell division factor.

Synonym(s):

6-Benzylaminopurine, Benzyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5
CAS Number:
Molecular Weight:
225.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

2 M NaOH: 40 mg/mL
ethanol: 5 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C12H11N5/c1-2-4-9(5-3-1)6-13-11-10-12(15-7-14-10)17-8-16-11/h1-5,7-8H,6H2,(H2,13,14,15,16,17)

InChI key

NWBJYWHLCVSVIJ-UHFFFAOYSA-N

General description

Plant cell division factor. Stimulates Ca2+ influx (Km = 1 nM) by moss protoplasts. Reported to induce positive inotropic effects through activation of P2-purinergic receptors. When complexed with copper, catalyzes molecular oxygen dismutation at physiological pH.
Plant cell division factor. Stimulates Ca2+ influx (Km = 1 nM) by moss protoplasts. When complexed with copper, catalyzes molecular oxygen dismutation at physiological pH.

Biochem/physiol Actions

Cell permeable: no
Km = 1 nM in stimulating Ca2+ influx
Primary Target
P2-purinergic receptors
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Froldi, G., et al. 1999. Planta Med.65, 245.
Schumaker, K.S., and Gizinski, M.J. 1993. Proc. Natl. Acad. Sci. USA 90, 10937.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service