Skip to Content
Merck
All Photos(1)

Documents

1.46419

Millipore

Buffered NaCl-Peptone Solution

ready-to-use, bottle volume 200 mL , filling volume

Synonym(s):

Buffered Saline Peptone Water, Buffered Sodium Chloride Peptone Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP 2.6.13

Quality Level

description

For the microbiological examination of non-sterile pharmaceuticals

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

single packed of 200 mL (in 250 ml bottle with blue screw cap and 3 loci (6 bottles per box))
bottle of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

250 mL

bottle volume

200 mL , filling volume

pH

7.0

application(s)

cosmetics

storage temp.

2-25°C

General description

Buffered NaCl-Peptone solution is designed for the microbial examination of non-sterile pharmaceutical products. The low content of peptone, along with phosphate buffer and sodium chloride in the solution maintains the viability of microorganisms present without supporting their growth.

Application

Buffered NaCl-Peptone solution is used for the microbial examination of non-sterile pharmaceutical products such as solutions for sample preparation, dilution, or washing solution.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Our media are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service