Skip to Content
Merck
All Photos(1)

Documents

1.13898

Supelco

Standard oil

for atomic absorption spectroscopy

Synonym(s):

Standard oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12181601
EC Index Number:
232-455-8
NACRES:
NA.24

grade

reference material

Quality Level

vapor pressure

0.0001 hPa ( 20 °C)

autoignition temp.

>300 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

kinematic viscosity

16 cSt(40 °C)

transition temp

flash point >180 °C

density

0.85 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

Analysis Note

Identity (IR-spectrum): passes test
Density (d 20 °C/ 4 °C): 0.84 - 0.86 g/ml
Viscosity (20 °C): 30 - 40 mPa∙s
Ag (Silver): ≤ 0.5 ppm
Al (Aluminium): ≤ 0.5 ppm
Au (Gold): ≤ 0.5 ppm
B (Boron): ≤ 0.5 ppm
Ba (Barium): ≤ 0.5 ppm
Be (Beryllium): ≤ 0.5 ppm
Bi (Bismuth): ≤ 0.5 ppm
Ca (Calcium): ≤ 0.5 ppm
Cd (Cadmium): ≤ 0.5 ppm
Co (Cobalt): ≤ 0.5 ppm
Cr (Chromium): ≤ 0.5 ppm
Cu (Copper): ≤ 0.5 ppm
Fe (Iron): ≤ 0.5 ppm
Ga (Gallium): ≤ 0.5 ppm
In (Indium): ≤ 0.5 ppm
K (Potassium): ≤ 0.5 ppm
Li (Lithium): ≤ 0.5 ppm
Mg (Magnesium): ≤ 0.5 ppm
Mn (Manganese): ≤ 0.5 ppm
Mo (Molybdenum): ≤ 0.5 ppm
Na (Sodium): ≤ 0.5 ppm
Ni (Nickel): ≤ 0.5 ppm
Pb (Lead): ≤ 0.5 ppm
Pt (Platinum): ≤ 0.5 ppm
Sb (Antimony): ≤ 0.5 ppm
Sn (Tin): ≤ 0.5 ppm
Sr (Strontium): ≤ 0.5 ppm
Ti (Titanium): ≤ 0.5 ppm
Tl (Thallium): ≤ 0.5 ppm
V (Vanadium): ≤ 0.5 ppm
Zn (Zinc): ≤ 0.5 ppm
Zr (Zirconium): ≤ 0.5 ppm
Water: ≤ 0.01 %

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

275.0 °F

Flash Point(C)

135 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service