Skip to Content
Merck
All Photos(1)

Documents

1.07593

Supelco

Resorcinol

GR for analysis

Synonym(s):

Resorcinol, 1,3-Dihydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OH)2
CAS Number:
Molecular Weight:
110.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-585-2
NACRES:
NA.21

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

605 °C

potency

301 mg/kg LD50, oral (Rat)
3360 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4 % (v/v)

pH

4-6 (20 °C, 100 g/L in H2O)

bp

281 °C/1013 hPa

mp

109-111 °C

transition temp

flash point 127 °C

solubility

1000 g/L

density

1.28 g/cm3 at 20 °C

bulk density

600‑700 kg/m3

storage temp.

15-25°C

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (bromatometric, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Melting range (lower value): ≥ 109.0 °C
Melting range (upper value): ≤ 111.0 °C
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Catechol: passes test
Phenol: ≤ 0.01 %
Sulfated ash: ≤ 0.02 %
Loss on drying: ≤ 1.0 %
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service