Skip to Content
Merck
All Photos(1)

Documents

1.04179

Sigma-Aldrich

Brilliant cresyl blue zinc chloride double salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H20N3O)2 · ZnCl4
CAS Number:
Molecular Weight:
771.92
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21

form

solid

Quality Level

composition

Dye, ≥60% spectrophotometric assay

pH

5.5 (20 °C, 5 g/L in H2O)

mp

195-198 °C

solubility

30 g/L

bulk density

560 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/2C17H20N3O.4ClH.Zn/c2*1-4-20(5-2)12-6-7-14-16(9-12)21-17-10-13(18)11(3)8-15(17)19-14;;;;;/h2*6-10H,4-5,18H2,1-3H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

SIHASHFMDFXGSX-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Brilliant cresyl blue zinc chloride double salt for microscopy Certistain®, is a dry staining dye that is used to prepare a staining solution, that makes target structures (by staining with the above brilliant cresyl blue solution) evaluable for diagnostic purposes. The regenerative capacity of erythrocytes can be detected by counting of reticulocytes. The measurement of substantia granulofilamentosa (ribonucleoproteins) is possible with fresh, non-fixed, young erythrocytes (supravital staining).Four stages of substantia granulofilamentosa maturation can be distinguished depending on the stage of reticulocyte development: coiled skein (I), incomplete network (II), complete network (III) and granular form (IV). In peripheral blood the development stages III and IV are found most commonly. When stained with brilliant cresyl blue the reticulocytes display a black-blue network or black-blue dots. As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

Dye content (spectrophotometrically): ≥ 60% C₁₇H₂₀N₃O
Identity
- UV/VIS-Spectrum:
passes test
- Zinc: passes test
Absorption maximum λ max (ethanol 50 %): 623 - 628 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; ethanol 50 %): ≥ 1500
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service