Skip to Content
Merck
All Photos(1)

Documents

1.03087

Supelco

1,5-Diphenylcarbazone

(cont. 50% Diphenylcarbazid) ACS,Reag. Ph Eur

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NB.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

mp

153-158 °C (decomposition)

bulk density

380 kg/m3

storage temp.

15-25°C

Application


  • Dosimetric characterization of a Nickel complex for routine use in radiation processing: This study explores the use of 1,5-Diphenylcarbazone in the dosimetric characterization of nickel complexes. This is particularly significant for researchers and scientists in radiation processing and material science, emphasizing its utility in radiation dosimetry and safety assessments (Babaei et al., 2022).

  • Determination of copper in traditional coffee pot water samples by flame atomic absorption spectrometry: This research utilizes 1,5-Diphenylcarbazone in a novel method for the detection of copper in water samples, showcasing its application in environmental monitoring and heavy metal detection. This finding is crucial for environmental lab technicians and researchers focusing on the accuracy and efficiency of metal detection in environmental samples (Ercan et al., 2020).

  • Hybrid mesoporous materials for on-line preconcentration of Cr(VI) followed by one-step scheme for elution and colorimetric determination at ultratrace levels: This study highlights the role of 1,5-Diphenylcarbazone in the development of hybrid mesoporous materials for the preconcentration and detection of chromium. It underscores the compound′s significant role in enhancing the sensitivity and specificity of environmental assays, which is invaluable for researchers engaged in the study of environmental contaminants and their mitigation (Kim et al., 2009).

Analysis Note

Purity (HPLC): 35 - 40
Identity (IR-spectrum): passes test
Solubility (Acetone): passes test
substances insoluble in ethanol: passes test
Melting point: 153 - 158
Sulfated ash: ≤ 0.1 %
Suitability for mercury determination: passes test
Diphenylcarbazone compound with Diphenylcarbazide (1:1)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service