Skip to Content
Merck

1.02584

SAFC

Caffeine

EMPROVE® ESSENTIAL Ph Eur,BP,USP

Pharma Manufacturing

Synonym(s):

Caffeine, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-362-1

Agency

BP
Ph. Eur.
USP

Quality Level

vapor pressure

20 hPa ( 80 °C)

product line

EMPROVE® ESSENTIAL

Assay

98.5-101.0% (HPLC)
98.5-101.0% (perchloric acid titration)

form

crystalline powder

autoignition temp.

>600 °C

potency

367.7 mg/kg LD50, oral (Rat)

technique(s)

API processing | cocrystal formation: suitable

pH

5.5-6.5 (20 °C, 10 g/L in H2O)

mp

235-239 °C

solubility

20 g/L

density

1.23 g/cm3 at 18 °C

application(s)

pharmaceutical

storage temp.

2-30°C

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Caffeine Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Strat-M® synthetic membrane mimics the human skin for In-vitro permeation testing (IVPT) of topical and transdermal formulations

Strat-M® synthetic membrane mimics the human skin for In-vitro permeation testing (IVPT) of topical and transdermal formulations

Strat-M® synthetic membrane mimics the human skin for In-vitro permeation testing (IVPT) of topical and transdermal formulations

Strat-M® synthetic membrane mimics the human skin for In-vitro permeation testing (IVPT) of topical and transdermal formulations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service